Assay Details
| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF072248.1 | - | 6 - 7 | 1108 | 133 | |
| BC038264.1 | - | 6 - 7 | 909 | 5250237 | 133 | |
| BP770230.1 | - | 1 - 2 | 133 | |||
| BQ042794.1 | - | 5 - 6 | 676 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282968 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282966 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282961 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282967 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282963 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282964 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282937 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960873 | BC038264.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960873 | NM_013595.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960873 | AF072248.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P127024 | BC038264.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P127024 | NM_013595.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P127024 | AF072248.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576123 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46448 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129622 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P127024 | BC038264.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P127024 | NM_013595.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P127024 | AF072248.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960873 | BC038264.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960873 | NM_013595.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960873 | AF072248.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P398004 | BC038264.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P398004 | NM_013595.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P398004 | AF072248.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282950 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129622 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02168883 | - | - |
| Whole Mouse Genome G4122A | A_51_P398004 | BC038264.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P398004 | NM_013595.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P398004 | AF072248.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122392 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127024 | BC038264.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127024 | NM_013595.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127024 | AF072248.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129622 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08583820 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08583824 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08741487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282964 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282937 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282966 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282967 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282963 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282961 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27283007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08846608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08901978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08367509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08629821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08367505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08506426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08886667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08291953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08380502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282987 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10370832 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370826 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370827 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370831 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370828 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370830 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417728_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10370824 | - | - |
| Mouse Genome 430 2.0 Array | 1417728_at | - | - |
| Mouse Genome 430 2.0 Array | 1417728_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17192_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17192_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000018484.mm.1 | NM_013595.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018484.mm.1 | TC1000002499.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018485.mm.1 | NM_013595.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018485.mm.1 | TC1000002499.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018483.mm.1 | NM_013595.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018483.mm.1 | TC1000002499.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417728_PM_at | - | - |
| Mouse Expression Set 430 | 1417728_at | - | - |
| Murine Genome U74v2 Set | 101385_PM_at | - | - |
| Murine Genome U74v2 Set | 101385_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417728_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417728_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417728_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417728_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6130364 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6130364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006130364 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2730498 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2730498 | - | - |
| mouse-6 v1.1 (Illumina) | scl37735.5.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6130364 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37735.5.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130364 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2730498 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2730498 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37735.5.1_7-S | - | - |

Mbd3
Genomic Map