Assay Details
| Gene Symbol: Msh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF107352.1 | - | 5 - 6 | 456 | 76 | |
| AF146227.1 | - | 4 - 5 | 664 | 76 | ||
| AK042083.1 | - | 5 - 6 | 441 | 76 | ||
| BC030034.1 | - | 3 - 4 | 744 | 76 | ||
| BC141113.1 | - | 4 - 5 | 584 | 9055725 | 76 | |
| BC145406.1 | - | 4 - 5 | 584 | 76 | ||
| BC145407.1 | - | 4 - 5 | 584 | 76 | ||
| BQ556345.1 | - | 5 - 6 | 453 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msh5
Genomic Map