Assay Details
| Gene Symbol: Msh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF107352.1 | - | 9 - 10 | 799 | 64 | |
| AF146227.1 | - | 8 - 9 | 1007 | 64 | ||
| AK042083.1 | - | 8 - 9 | 748 | 64 | ||
| BC141113.1 | - | 8 - 9 | 927 | 9055725 | 64 | |
| BC145406.1 | - | 8 - 9 | 927 | 64 | ||
| BC145407.1 | - | 8 - 9 | 927 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msh5
Genomic Map