Assay Details
| Gene Symbol: Msh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF107352.1 | - | 16 - 17 | 1439 | 71 | |
| AF146227.1 | - | 15 - 16 | 1647 | 71 | ||
| BC141113.1 | - | 15 - 16 | 1567 | 9055725 | 71 | |
| BC145407.1 | - | 15 - 16 | 1567 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msh5
Genomic Map