Assay Details
| Gene Symbol: Msh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF107352.1 | - | 21 - 22 | 2071 | 72 | |
| AF146227.1 | - | 20 - 21 | 2279 | 72 | ||
| AK132626.1 | - | 8 - 9 | 1140 | 72 | ||
| BC141113.1 | - | 20 - 21 | 2199 | 9055725 | 72 | |
| BC145406.1 | - | 20 - 21 | 2189 | 72 | ||
| BC145407.1 | - | 20 - 21 | 2101 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msh5
Genomic Map