Assay Details
| Gene Symbol: Syn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013722.3 | NP_038750.2 | 7 - 8 | 1020 | 72 | |
| GenBank mRNA | AF192754.1 | - | 7 - 8 | 965 | 72 | |
| AF498252.1 | - | 6 - 7 | 767 | 72 | ||
| AF498253.1 | - | 6 - 7 | 767 | 72 | ||
| AK048643.1 | - | 7 - 8 | 1058 | 72 | ||
| AK080557.1 | - | 7 - 8 | 73 | |||
| BC107397.2 | - | 6 - 7 | 886 | 40058225 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syn3
Genomic Map