Assay Details
| Gene Symbol: Clnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013748.3 | NP_038776.3 | 8 - 9 | 581 | 79 | |
| GenBank mRNA | AB021220.1 | - | 8 - 9 | 701 | 79 | |
| AF187819.1 | - | 8 - 9 | 581 | 79 | ||
| AK082826.1 | - | 4 - 5 | 319 | 79 | ||
| BC125637.1 | - | 8 - 9 | 578 | 40130152 | 79 | |
| BC125639.1 | - | 8 - 9 | 578 | 40130154 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clnk
Genomic Map