Assay Details
| Gene Symbol: Tnfrsf12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013749.2 | NP_038777.2 | 1 - 2 | 129 | 77 | |
| GenBank mRNA | AF156164.1 | - | 1 - 2 | 124 | 77 | |
| AK005382.1 | - | 1 - 2 | 126 | 77 | ||
| AK005530.1 | - | 1 - 2 | 125 | 77 | ||
| AK160136.1 | - | 1 - 2 | 130 | 77 | ||
| BC025860.1 | - | 1 - 2 | 113 | 5251646 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnfrsf12a
Genomic Map