Assay Details
| Gene Symbol: Msrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013759.2 | NP_038787.1 | 3 - 4 | 387 | 80 | |
| GenBank mRNA | AF195142.1 | - | 3 - 4 | 362 | 80 | |
| AK002339.1 | - | 3 - 4 | 347 | 80 | ||
| AK002652.1 | - | 3 - 4 | 346 | 80 | ||
| AK003624.1 | - | 3 - 4 | 347 | 80 | ||
| BC090646.1 | - | 3 - 4 | 387 | 30946285 | 80 | |
| BC141146.1 | - | 3 - 4 | 351 | 9055758 | 80 | |
| CX566836.1 | - | 3 - 4 | 387 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msrb1
Genomic Map