Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
TC0500001093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NM_001253877.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NM_001253877.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NM_001253878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NM_001253878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NM_001253879.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NM_001253879.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NM_001253880.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NM_001253880.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NM_013827.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NM_013827.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NR_045608.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009253.mm.1 |
NR_045608.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
TC0500001093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NM_001253877.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NM_001253877.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NM_001253878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NM_001253878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NM_001253879.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NM_001253879.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NM_001253880.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NM_001253880.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NM_013827.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NM_013827.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NR_045608.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009254.mm.1 |
NR_045608.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
TC0500001093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NM_001253877.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NM_001253877.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NM_001253878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NM_001253878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NM_001253879.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NM_001253879.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NM_001253880.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NM_001253880.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NM_013827.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NM_013827.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NR_045608.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009255.mm.1 |
NR_045608.1 |
Refseq |