Assay Details
| Gene Symbol: Cabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013877.3 | NP_038905.1 | 3 - 4 | 390 | 74 | |
| GenBank mRNA | AB079605.1 | - | 2 - 3 | 206 | 74 | |
| AF169161.1 | - | 3 - 4 | 351 | 74 | ||
| AK044544.1 | - | 3 - 4 | 391 | 74 | ||
| AK087416.1 | - | 3 - 4 | 251 | 74 | ||
| BC018253.1 | - | 3 - 4 | 413 | 4504283 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cabp5
Genomic Map