Assay Details
| Gene Symbol: Cngb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013927.2 | NP_038955.1 | 12 - 13 | 1535 | 67 | |
| GenBank mRNA | AJ243572.1 | - | 12 - 13 | 1535 | 67 | |
| AK040140.1 | - | 3 - 4 | 402 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cngb3
Genomic Map