Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200015389.mm.1 |
NM_001159593.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015389.mm.1 |
NM_001159593.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015389.mm.1 |
NM_015747.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015389.mm.1 |
NM_015747.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015389.mm.1 |
TC0200001951.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200015387.mm.1 |
NM_001159593.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015387.mm.1 |
NM_001159593.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015387.mm.1 |
NM_015747.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015387.mm.1 |
NM_015747.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015387.mm.1 |
TC0200001951.mm.1 |
MTA TC |