Assay Details
| Gene Symbol: Sufu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025391.2 | NP_001020562.1 | 4 - 5 | 798 | 69 | |
| NM_015752.3 | NP_056567.2 | 4 - 5 | 798 | 69 | ||
| GenBank mRNA | AF134893.1 | - | 4 - 5 | 738 | 69 | |
| AJ131692.1 | - | 4 - 5 | 799 | 69 | ||
| AJ308625.1 | - | 4 - 5 | 477 | 69 | ||
| AJ308626.1 | - | 4 - 5 | 732 | 69 | ||
| AK015885.1 | - | 3 - 4 | 556 | 69 | ||
| AK047603.1 | - | 4 - 5 | 668 | 69 | ||
| AK156410.1 | - | 4 - 5 | 760 | 69 | ||
| BC048168.1 | - | 4 - 5 | 771 | 6821477 | 69 | |
| BC056997.1 | - | 4 - 5 | 771 | 6821477 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sufu
Genomic Map