Assay Details
| Gene Symbol: Sh3bgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015825.2 | NP_056640.1 | 2 - 3 | 464 | 122 | |
| GenBank mRNA | AJ239082.1 | - | 2 - 3 | 230 | 122 | |
| AJ272170.1 | - | 2 - 3 | 292 | 122 | ||
| AJ487014.1 | - | 2 - 3 | 291 | 122 | ||
| BC055870.1 | - | 2 - 3 | 281 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033135 | AJ487014.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033135 | BC055870.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033135 | NM_015825.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952915 | AJ487014.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952915 | BC055870.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952915 | NM_015825.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300285 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311194 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 220721 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52720 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805133 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921710 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451655 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109576 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477422 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599068 | M63697.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 220514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93430 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4952 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117646 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177363 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033135 | AJ487014.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033135 | BC055870.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033135 | NM_015825.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952915 | AJ487014.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952915 | BC055870.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952915 | NM_015825.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127718 | AJ487014.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127718 | BC055870.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127718 | NM_015825.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127718 | AJ239082.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P127718 | AJ272170.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991582 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08694981 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08694982 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117646 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08694941 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08694953 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08694971 | - | - |
| Whole Mouse Genome G4122A | A_51_P127718 | AJ487014.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127718 | BC055870.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127718 | NM_015825.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127718 | AJ239082.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P127718 | AJ272170.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114790 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08694982 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117646 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08694941 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08694981 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12495180 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12495175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12995249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12500348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12989338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12495183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12500350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12495179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12500354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12570962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12585841 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10437186 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437182 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437183 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437181 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422644_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454559_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433319_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10437180 | - | - |
| Mouse Genome 430 2.0 Array | 1422644_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422644_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50795_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50795_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600008541.mm.1 | NM_015825.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008541.mm.1 | NM_015825.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008541.mm.1 | TC1600001080.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008542.mm.1 | NM_015825.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008542.mm.1 | NM_015825.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008542.mm.1 | TC1600001080.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008548.mm.1 | NM_015825.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008548.mm.1 | NM_015825.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008548.mm.1 | TC1600001080.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008549.mm.1 | NM_015825.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008549.mm.1 | NM_015825.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008549.mm.1 | TC1600001080.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008550.mm.1 | NM_015825.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008550.mm.1 | NM_015825.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008550.mm.1 | TC1600001080.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422644_at | - | - |
| Mouse Expression Set 430 | 1422644_PM_at | - | - |
| Murine Genome U74v2 Set | 161935_r_at | - | - |
| Murine Genome U74v2 Set | 96205_at | - | - |
| Murine Genome U74v2 Set | 161935_PM_r_at | - | - |
| Murine Genome U74v2 Set | 96204_PM_at | - | - |
| Murine Genome U74v2 Set | 96204_at | - | - |
| Murine Genome U74v2 Set | 96205_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422644_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422644_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422644_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422644_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433319_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454559_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433319_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454559_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 792871 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2679128 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2679128 | - | - |

Sh3bgr
Genomic Map