Assay Details
| Gene Symbol: Slc25a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177572.1 | NP_001171043.1 | 10 - 11 | 1179 | 89 | |
| NM_015829.3 | NP_056644.1 | 10 - 11 | 1182 | 89 | ||
| GenBank mRNA | AF164632.1 | - | 10 - 11 | 1127 | 89 | |
| AK002829.1 | - | 10 - 11 | 1057 | 89 | ||
| AK012670.1 | - | 10 - 11 | 1070 | 89 | ||
| AK029020.1 | - | 10 - 11 | 1110 | 89 | ||
| AK150496.1 | - | 10 - 11 | 1125 | 89 | ||
| AK151180.1 | - | 10 - 11 | 1125 | 89 | ||
| AK152237.1 | - | 10 - 11 | 1125 | 89 | ||
| BC016571.1 | - | 4 - 5 | 316 | 3600909 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a13
Genomic Map