Assay Details
| Gene Symbol: Capn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015830.1 | NP_056645.1 | 3 - 4 | 1875 | 63 | |
| GenBank mRNA | AK143711.1 | - | 5 - 6 | 2514 | 63 | |
| AK148209.1 | - | 1 - 2 | 134 | 64 | ||
| AK170543.1 | - | 4 - 5 | 2109 | 63 | ||
| BC058094.1 | - | 4 - 5 | 2133 | 6824892 | 63 | |
More Information
Set Membership: |
|

Capn15
Genomic Map