Assay Details
| Gene Symbol: Galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK134525.1 | - | 1 - 2 | 154 | 52 | |
| AK163544.1 | - | 1 - 2 | 185 | 52 | ||
| BY289084.1 | - | 3 - 4 | 375 | 52 | ||
| CX239267.1 | - | 3 - 4 | 367 | 52 | ||
| M96265.1 | - | 3 - 4 | 401 | 52 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK163544.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22864173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22864172 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22864171 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22864174 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149269 | NM_016658.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149269 | AK134525.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2066927 | AK088379.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73484 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118767 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149269 | NM_016658.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149269 | AK134525.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P117777 | M96265.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P117777 | NM_016658.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P117777 | AK134525.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P117777 | AK163544.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P117777 | AV027352.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22864169 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118767 | - | - |
| Whole Mouse Genome G4122A | A_51_P117777 | M96265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117777 | NM_016658.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117777 | AK134525.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117777 | AK163544.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117777 | AV027352.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137552 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118581 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118767 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03492089 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03492107 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03492091 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03492101 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03492102 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03492104 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03736448 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03736449 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22864174 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22864171 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22864172 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22864173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22864221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03930105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04022735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04022733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04022732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04022730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03736448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22864141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03492091 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10504096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10504097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10504099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10504098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10504100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10504101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10504102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10504103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10504095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10504104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10504105 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450187_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10504094 | - | - |
| Mouse Genome 430 2.0 Array | 1450187_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450187_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14430_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14430_at | - | - |
| Mouse Expression Set 430 | 1450187_a_at | - | - |
| Mouse Expression Set 430 | 1450187_PM_a_at | - | - |
| Murine Genome U74v2 Set | 104616_g_at | - | - |
| Murine Genome U74v2 Set | 104615_at | - | - |
| Murine Genome U74v2 Set | 104616_PM_g_at | - | - |
| Murine Genome U74v2 Set | 104615_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450187_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450187_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450187_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450187_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5550075 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3710162 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3710162 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5550075 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003710162 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550075 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2711442 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2677567 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2711442 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2677567 | - | - |
| mouse-6 v1.1 (Illumina) | scl25528.7.1_41-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0002692.1_59-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3710162 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550075 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25528.7.1_41-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002692.1_59-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550075 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710162 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2711442 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2677567 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2711442 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2677567 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25528.7.1_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0002692.1_59-S | - | - |

Galt
Genomic Map