Assay Details
| Gene Symbol: Mpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016695.3 | NP_057904.1 | 7 - 8 | 935 | 71 | |
| GenBank mRNA | AF162685.1 | - | 7 - 8 | 922 | 71 | |
| AK141422.1 | - | 6 - 7 | 933 | 71 | ||
| AK161577.1 | - | 7 - 8 | 928 | 71 | ||
| AK172253.1 | - | 7 - 8 | 920 | 71 | ||
| AK188703.1 | - | 2 - 3 | 216 | 71 | ||
| BC053026.1 | - | 7 - 8 | 935 | 5709543 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpp2
Genomic Map