Assay Details
| Gene Symbol: Scly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016717.3 | NP_057926.2 | 5 - 6 | 637 | 85 | |
| GenBank mRNA | AF175407.1 | - | 5 - 6 | 589 | 85 | |
| AK036533.1 | - | 5 - 6 | 606 | 85 | ||
| AK036750.1 | - | 5 - 6 | 639 | 85 | ||
| AK186846.1 | - | 1 - 2 | 71 | 85 | ||
| BC019879.1 | - | 5 - 6 | 597 | 5123843 | 85 | |
| BC021389.1 | - | 5 - 6 | 621 | 5054691 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scly
Genomic Map