Assay Details
| Gene Symbol: Stip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016737.2 | NP_058017.1 | 1 - 2 | 143 | 82 | |
| GenBank mRNA | AK088494.1 | - | 1 - 2 | 85 | 82 | |
| AK134171.1 | - | 1 - 2 | 59 | 82 | ||
| AK149493.1 | - | 1 - 2 | 59 | 82 | ||
| AK161645.1 | - | 1 - 2 | 62 | 82 | ||
| AK167273.1 | - | 1 - 2 | 77 | 82 | ||
| AK168181.1 | - | 1 - 2 | 59 | 82 | ||
| BC003794.1 | - | 1 - 2 | 52 | 3590457 | 82 | |
| CJ155828.1 | - | 1 - 2 | 144 | 82 | ||
| U27830.1 | - | 1 - 2 | 60 | 82 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK088494.1,AK149493.1,AK161645.1,AK167273.1,AK168181.1,BC003794.1,NM_016737.2,U27830.1 transcripts.
Set Membership: |
|

Stip1
Genomic Map