Assay Details
| Gene Symbol: Cdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016742.4 | NP_058022.1 | 7 - 8 | 1036 | 74 | |
| GenBank mRNA | AK010494.1 | - | 7 - 8 | 1061 | 74 | |
| AK013255.1 | - | 7 - 8 | 1037 | 74 | ||
| AK145671.1 | - | 7 - 8 | 1068 | 74 | ||
| AK168651.1 | - | 7 - 8 | 1054 | 74 | ||
| BC060079.1 | - | 7 - 8 | 1059 | 5714770 | 74 | |
| U43076.1 | - | 7 - 8 | 1074 | 74 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
posttranscriptional regulation of gene expression
regulation of protein kinase activity
protein stabilization
regulation of interferon-gamma-mediated signaling pathway
regulation of type I interferon-mediated signaling pathway
mitophagy in response to mitochondrial depolarization
protein C-terminus binding
protein kinase regulator activity
kinase binding
protein kinase binding
heat shock protein binding
mitogen-activated protein kinase kinase kinase binding
protein kinase B binding
unfolded protein binding
chaperone binding
Hsp90 protein binding
protein folding
posttranscriptional regulation of gene expression
regulation of protein kinase activity
protein stabilization
regulation of interferon-gamma-mediated signaling pathway
regulation of type I interferon-mediated signaling pathway
mitophagy in response to mitochondrial depolarization
protein C-terminus binding
protein kinase regulator activity
kinase binding
protein kinase binding
heat shock protein binding
mitogen-activated protein kinase kinase kinase binding
protein kinase B binding
unfolded protein binding
chaperone binding
Hsp90 protein binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345696 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345698 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345697 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345695 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P463143 | AK168651.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P463143 | BC060079.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P463143 | AK013255.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P463143 | AK145671.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P463143 | NM_016742.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P463143 | AK168651.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P463143 | BC060079.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P463143 | AK013255.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P463143 | AK145671.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P463143 | NM_016742.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906701 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 196363 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 273941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 836248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201203 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59683 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173654 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135031 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P463143 | AK168651.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P463143 | BC060079.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P463143 | AK013255.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P463143 | AK145671.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P463143 | NM_016742.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P463143 | AK168651.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P463143 | BC060079.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P463143 | AK013255.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P463143 | AK145671.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P463143 | NM_016742.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345688 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01881629 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135031 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173654 | - | - |
| Whole Mouse Genome G4122A | A_51_P425749 | AK168651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P425749 | BC060079.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P425749 | U43076.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P425749 | AK013255.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P425749 | AK145671.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463143 | AK168651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463143 | BC060079.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463143 | AK013255.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463143 | AK145671.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463143 | NM_016742.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463143 | AK168651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463143 | BC060079.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463143 | AK013255.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463143 | AK145671.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P463143 | NM_016742.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121689 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173654 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135031 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08161031 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08161030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345696 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345695 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345698 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08010615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08010619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08015845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08198754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08161030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08161031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07533908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07468892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08006467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08006472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07922129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08163313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07922139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08006471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07922133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345723 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591479 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591480 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591477 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591478 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591476 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591473 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591475 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591481 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591474 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416819_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416820_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591472 | - | - |
| Mouse Genome 430 2.0 Array | 1416820_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416819_at | - | - |
| Mouse Genome 430 2.0 Array | 1416820_at | - | - |
| Mouse Genome 430 2.0 Array | 1416819_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12539_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12539_at | - | - |
| Mouse Expression Set 430 | 1416820_PM_at | - | - |
| Mouse Expression Set 430 | 1416819_at | - | - |
| Mouse Expression Set 430 | 1416819_PM_at | - | - |
| Mouse Expression Set 430 | 1416820_at | - | - |
| Murine Genome U74v2 Set | 99135_at | - | - |
| Murine Genome U74v2 Set | 99135_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416819_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416820_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416820_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416819_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416820_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416819_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416819_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416820_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2570537 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002570537 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2721966 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2721966 | - | - |
| mouse-6 v1.1 (Illumina) | scl36151.10_243-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2570537 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36151.10_243-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2570537 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2721966 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2721966 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36151.10_243-S | - | - |

Cdc37
Genomic Map