Assay Details
| Gene Symbol: Smad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016769.4 | NP_058049.3 | 4 - 5 | 925 | 96 | |
| GenBank mRNA | AB008192.1 | - | 4 - 5 | 634 | 96 | |
| AF016189.1 | - | 4 - 5 | 901 | 96 | ||
| AK041743.1 | - | 4 - 5 | 446 | 96 | ||
| AK048626.1 | - | 4 - 5 | 925 | 96 | ||
| AK083158.1 | - | 4 - 5 | 533 | 96 | ||
| BC066850.1 | - | 4 - 5 | 611 | 30432720 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smad3
Genomic Map