Assay Details
| Gene Symbol: Cpsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016856.3 | NP_058552.1 | 5 - 6 | 744 | 68 | |
| GenBank mRNA | AF012822.1 | - | 5 - 6 | 730 | 68 | |
| AK054372.1 | - | 5 - 6 | 727 | 68 | ||
| AK140869.1 | - | 5 - 6 | 726 | 68 | ||
| AK141325.1 | - | 5 - 6 | 726 | 68 | ||
| AK147746.1 | - | 5 - 6 | 745 | 68 | ||
| BC007163.1 | - | 3 - 4 | 402 | 68 | ||
| BC013628.1 | - | 5 - 6 | 719 | 3497854 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf2
Genomic Map