Assay Details
| Gene Symbol: Fut8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252614.1 | NP_001239543.1 | 6 - 7 | 928 | 139 | |
| NM_016893.5 | NP_058589.2 | 5 - 6 | 839 | 139 | ||
| NR_045554.1 | 5 - 6 | 1258 | 139 | |||
| GenBank mRNA | AB025198.1 | - | 2 - 3 | 528 | 139 | |
| AK048520.1 | - | 5 - 6 | 977 | 139 | ||
| AK051811.1 | - | 5 - 6 | 796 | 139 | ||
| AK134537.1 | - | 5 - 6 | 932 | 139 | ||
| AK163494.1 | - | 5 - 6 | 955 | 139 | ||
| BC010666.1 | - | 6 - 7 | 907 | 3594582 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fut8
Genomic Map