Assay Details
| Gene Symbol: Cdkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016912.2 | NP_058608.1 | 12 - 13 | 1915 | 74 | |
| GenBank mRNA | AB029065.1 | - | 12 - 13 | 1891 | 74 | |
| AK030598.1 | - | 12 - 13 | 2170 | 74 | ||
| AK144574.1 | - | 12 - 13 | 1916 | 74 | ||
| BC117894.1 | - | 12 - 13 | 1824 | 40096852 | 74 | |
| BC117895.1 | - | 12 - 13 | 1824 | 40096853 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdkl2
Genomic Map