Assay Details
| Gene Symbol: Cdkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016912.2 | NP_058608.1 | 13 - 14 | 2004 | 75 | |
| GenBank mRNA | AB029065.1 | - | 13 - 14 | 1980 | 75 | |
| AK030598.1 | - | 13 - 14 | 2259 | 75 | ||
| AK144574.1 | - | 13 - 14 | 2005 | 75 | ||
| BC117894.1 | - | 13 - 14 | 1913 | 40096852 | 75 | |
| BC117895.1 | - | 13 - 14 | 1913 | 40096853 | 75 | |
More Information
Set Membership: |
|

Cdkl2
Genomic Map