Assay Details
| Gene Symbol: Slc40a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016917.2 | NP_058613.2 | 1 - 2 | 381 | 80 | |
| GenBank mRNA | AF215637.1 | - | 1 - 2 | 334 | 80 | |
| AF226613.1 | - | 1 - 2 | 347 | 80 | ||
| AF231120.1 | - | 1 - 2 | 320 | 80 | ||
| AK008700.1 | - | 1 - 2 | 185 | 80 | ||
| AK033987.1 | - | 1 - 2 | 397 | 80 | ||
| AK144780.1 | - | 1 - 2 | 368 | 80 | ||
| AK147137.1 | - | 1 - 2 | 367 | 80 | ||
| AK159294.1 | - | 1 - 2 | 370 | 80 | ||
| AK159855.1 | - | 1 - 2 | 367 | 80 | ||
| BB625854.1 | - | 1 - 2 | 397 | 80 | ||
| BC003438.1 | - | 1 - 2 | 350 | 2647365 | 80 | |
| FJ207421.1 | - | 2 - 3 | 291 | 80 | ||
| FJ207422.1 | - | 2 - 3 | 248 | 80 | ||
| FJ207423.1 | - | 2 - 3 | 210 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc40a1
Genomic Map