Assay Details
| Gene Symbol: Clpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017393.2 | NP_059089.1 | 5 - 6 | 715 | 97 | |
| GenBank mRNA | AJ005253.1 | - | 5 - 6 | 683 | 97 | |
| AK002238.1 | - | 5 - 6 | 654 | 97 | ||
| AK004024.1 | - | 5 - 6 | 664 | 97 | ||
| AK145765.1 | - | 5 - 6 | 677 | 97 | ||
| AK147719.1 | - | 5 - 6 | 643 | 97 | ||
| AK168053.1 | - | 5 - 6 | 662 | 97 | ||
| AK179220.1 | - | 2 - 3 | 284 | 97 | ||
| BC001998.1 | - | 5 - 6 | 674 | 3487489 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
misfolded or incompletely synthesized protein catabolic process
protein homooligomerization
proteolysis involved in cellular protein catabolic process
serine-type endopeptidase activity
protein binding
peptidase activity
serine-type peptidase activity
hydrolase activity
identical protein binding
proteolysis
misfolded or incompletely synthesized protein catabolic process
protein homooligomerization
proteolysis involved in cellular protein catabolic process
serine-type endopeptidase activity
protein binding
peptidase activity
serine-type peptidase activity
hydrolase activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259429 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134739 | BC001998.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134739 | AK168053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134739 | AK004024.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134739 | NM_017393.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134739 | AJ005253.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 271097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190626 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124508 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118674 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134739 | BC001998.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134739 | AK168053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134739 | AK004024.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134739 | NM_017393.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134739 | AJ005253.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139021 | AJ005253.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139021 | BC001998.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139021 | AK168053.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139021 | AK004024.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139021 | NM_017393.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31259455 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118674 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03340627 | - | - |
| Whole Mouse Genome G4122A | A_51_P139021 | AJ005253.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139021 | BC001998.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139021 | AK168053.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139021 | AK004024.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139021 | NM_017393.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134739 | BC001998.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134739 | AK168053.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134739 | AK004024.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134739 | NM_017393.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134739 | AJ005253.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118674 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13434760 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31259429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31259484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13421979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13434764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13434752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13162940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13294095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13456136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13423225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13200857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13456132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13425818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31259443 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10446212 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446209 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446213 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446208 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446210 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416616_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416615_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10446207 | - | - |
| Mouse Genome 430 2.0 Array | 1416615_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416616_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1416615_at | - | - |
| Mouse Genome 430 2.0 Array | 1416616_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53895_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53895_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700009843.mm.1 | NM_017393.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009843.mm.1 | NM_017393.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009843.mm.1 | TC1700001020.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700009844.mm.1 | NM_017393.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009844.mm.1 | NM_017393.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009844.mm.1 | TC1700001020.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416615_PM_at | - | - |
| Mouse Expression Set 430 | 1416615_at | - | - |
| Mouse Expression Set 430 | 1416616_PM_s_at | - | - |
| Mouse Expression Set 430 | 1416616_s_at | - | - |
| Murine Genome U74v2 Set | 93048_at | - | - |
| Murine Genome U74v2 Set | 93048_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416616_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416615_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416615_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416616_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416616_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416615_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416616_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416615_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 406804 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6520102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006520102 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2771009 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2771009 | - | - |
| mouse-6 v1.1 (Illumina) | scl50582.4.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6520102 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50582.4.1_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520102 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2982890 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2982890 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2771009 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50582.4.1_9-S | - | - |

Clpp
Genomic Map