Assay Details
| Gene Symbol: Rdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001150749.1 | NP_001144221.1 | 3 - 4 | 853 | 79 | |
| NM_017473.4 | NP_059501.1 | 3 - 4 | 825 | 79 | ||
| GenBank mRNA | AB015165.1 | - | 3 - 4 | 941 | 79 | |
| AB032058.1 | - | 3 - 4 | 733 | 79 | ||
| AF056194.1 | - | 3 - 4 | 830 | 79 | ||
| BC024603.1 | - | 3 - 4 | 803 | 4196075 | 79 | |
| BC092255.1 | - | 3 - 4 | 779 | 5100891 | 79 | |
| BC093514.1 | - | 3 - 4 | 767 | 5132290 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968399 | BC093514.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968399 | CB951082.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968399 | AF056194.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968399 | AI255893.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968399 | BC024603.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12833 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111257 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968399 | BC093514.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968399 | CB951082.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968399 | AF056194.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968399 | AI255893.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968399 | BC024603.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P122170 | BC093514.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P122170 | CB951082.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P122170 | AF056194.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P122170 | AI255893.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P122170 | BC024603.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538521 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07231555 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111257 | - | - |
| Whole Mouse Genome G4122A | A_51_P122170 | BC093514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122170 | CB951082.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122170 | AF056194.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122170 | AI255893.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122170 | BC024603.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137073 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111257 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08916118 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08546112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08546093 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08916121 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08546098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08448218 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08704921 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08916125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08546109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08546092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08504217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08546117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08851884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08451624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08546106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08851876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08451619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08546094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538529 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373331 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448723_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373330 | - | - |
| Mouse Genome 430 2.0 Array | 1448723_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448723_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54150_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54150_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000023459.mm.1 | NM_001150749.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023459.mm.1 | NM_001150749.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023459.mm.1 | NM_017473.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023459.mm.1 | NM_017473.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023459.mm.1 | TC1000003123.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448723_at | - | - |
| Mouse Expression Set 430 | 1448723_PM_at | - | - |
| Murine Genome U74v2 Set | 100634_at | - | - |
| Murine Genome U74v2 Set | 100634_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448723_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448723_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448723_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448723_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 928829 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 670739 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000670739 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226515 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226515 | - | - |
| mouse-6 v1.1 (Illumina) | scl37375.4.1_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 670739 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37375.4.1_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670739 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226515 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2871249 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226515 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2871249 | - | - |

Rdh7
Genomic Map