Assay Details
| Gene Symbol: Ctnnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018761.3 | NP_061231.3 | 9 - 10 | 1437 | 67 | |
| GenBank mRNA | AF006071.1 | - | 9 - 10 | 1424 | 67 | |
| AK034393.1 | - | 10 - 11 | 1437 | 67 | ||
| AK038548.1 | - | 6 - 7 | 1374 | 67 | ||
| AK082755.1 | - | 9 - 10 | 1381 | 67 | ||
| BC050070.1 | - | 9 - 10 | 1414 | 30012833 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnal1
Genomic Map