Assay Details
| Gene Symbol: Ctnnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018761.3 | NP_061231.3 | 11 - 12 | 1686 | 78 | |
| GenBank mRNA | AF006071.1 | - | 11 - 12 | 1673 | 78 | |
| AK082755.1 | - | 11 - 12 | 1630 | 78 | ||
| BC050070.1 | - | 11 - 12 | 1663 | 30012833 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnal1
Genomic Map