Assay Details
| Gene Symbol: Skap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018773.2 | NP_061243.1 | 10 - 11 | 1020 | 77 | |
| GenBank mRNA | AB014485.1 | - | 10 - 11 | 937 | 77 | |
| AF051324.1 | - | 10 - 11 | 935 | 77 | ||
| AK076000.1 | - | 10 - 11 | 965 | 77 | ||
| AK144289.1 | - | 10 - 11 | 957 | 77 | ||
| AK204408.1 | - | 2 - 3 | 156 | 77 | ||
| BC003711.1 | - | 10 - 11 | 1020 | 3599914 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Skap2
Genomic Map