| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200001435.mm.1 |
NM_018813.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001435.mm.1 |
NM_018813.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001435.mm.1 |
TC1200000181.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200001436.mm.1 |
NM_018813.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001436.mm.1 |
NM_018813.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001436.mm.1 |
TC1200000181.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200001433.mm.1 |
NM_018813.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001433.mm.1 |
NM_018813.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200001433.mm.1 |
TC1200000181.mm.1 |
MTA TC |