Assay Details
| Gene Symbol: Cpsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018813.3 | NP_061283.2 | 7 - 8 | 959 | 84 | |
| GenBank mRNA | AF203969.1 | - | 7 - 8 | 812 | 84 | |
| AK149953.1 | - | 7 - 8 | 886 | 84 | ||
| AK157205.1 | - | 7 - 8 | 901 | 84 | ||
| AK158036.1 | - | 7 - 8 | 871 | 84 | ||
| AK170842.1 | - | 7 - 8 | 844 | 84 | ||
| AK171838.1 | - | 7 - 8 | 890 | 84 | ||
| BC006748.1 | - | 5 - 6 | 612 | 84 | ||
| BC023297.1 | - | 7 - 8 | 845 | 4192224 | 84 | |
| BC100569.1 | - | 7 - 8 | 857 | 30041540 | 84 | |
| CF725936.1 | - | 5 - 6 | 568 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf3
Genomic Map