Assay Details
| Gene Symbol: Cpsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018813.3 | NP_061283.2 | 9 - 10 | 1295 | 91 | |
| GenBank mRNA | AF203969.1 | - | 9 - 10 | 1148 | 91 | |
| AK010566.1 | - | 2 - 3 | 362 | 91 | ||
| AK149953.1 | - | 9 - 10 | 1222 | 91 | ||
| AK157205.1 | - | 9 - 10 | 1237 | 91 | ||
| AK158036.1 | - | 9 - 10 | 1207 | 91 | ||
| AK170842.1 | - | 9 - 10 | 1180 | 91 | ||
| AK171838.1 | - | 9 - 10 | 1226 | 91 | ||
| BC023297.1 | - | 9 - 10 | 1181 | 4192224 | 91 | |
| BC100569.1 | - | 9 - 10 | 1193 | 30041540 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf3
Genomic Map