Assay Details
| Gene Symbol: Cpsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018813.3 | NP_061283.2 | 16 - 17 | 2053 | 87 | |
| GenBank mRNA | AF203969.1 | - | 16 - 17 | 1906 | 87 | |
| AK004317.1 | - | 3 - 4 | 263 | 87 | ||
| AK010566.1 | - | 9 - 10 | 1120 | 87 | ||
| AK045238.1 | - | 5 - 6 | 423 | 87 | ||
| AK149953.1 | - | 16 - 17 | 1980 | 87 | ||
| AK157205.1 | - | 16 - 17 | 1995 | 87 | ||
| AK158036.1 | - | 16 - 17 | 1966 | 87 | ||
| AK170842.1 | - | 16 - 17 | 1938 | 87 | ||
| AK171838.1 | - | 16 - 17 | 1984 | 87 | ||
| BC006748.1 | - | 11 - 12 | 1608 | 87 | ||
| BC023297.1 | - | 16 - 17 | 1939 | 4192224 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf3
Genomic Map