Assay Details
| Gene Symbol: Samhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139520.1 | NP_001132992.1 | 11 - 12 | 1444 | 84 | |
| NM_018851.3 | NP_061339.3 | 11 - 12 | 1444 | 84 | ||
| GenBank mRNA | AK054490.1 | - | 11 - 12 | 1343 | 84 | |
| AK078936.1 | - | 11 - 12 | 1387 | 84 | ||
| AK128954.1 | - | 10 - 11 | 1242 | 84 | ||
| AK151335.1 | - | 11 - 12 | 1318 | 84 | ||
| AK153390.1 | - | 11 - 12 | 1318 | 84 | ||
| AK170688.1 | - | 11 - 12 | 1343 | 84 | ||
| AK171565.1 | - | 11 - 12 | 1344 | 84 | ||
| BC012721.1 | - | 11 - 12 | 1381 | 4037046 | 84 | |
| BC067198.1 | - | 11 - 12 | 1367 | 6826682 | 84 | |
| U15635.1 | - | 11 - 12 | 1368 | 84 | ||
More Information
Set Membership: |
|

Samhd1
Genomic Map