Assay Details
| Gene Symbol: Samhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139520.1 | NP_001132992.1 | 13 - 14 | 1658 | 92 | |
| NM_018851.3 | NP_061339.3 | 13 - 14 | 1658 | 92 | ||
| GenBank mRNA | AK054490.1 | - | 13 - 14 | 1557 | 92 | |
| AK078936.1 | - | 13 - 14 | 92 | |||
| AK128954.1 | - | 12 - 13 | 1456 | 92 | ||
| AK151335.1 | - | 13 - 14 | 1532 | 92 | ||
| AK153390.1 | - | 13 - 14 | 1532 | 92 | ||
| AK170688.1 | - | 13 - 14 | 1557 | 92 | ||
| AK171565.1 | - | 13 - 14 | 1558 | 92 | ||
| BC012721.1 | - | 13 - 14 | 1595 | 4037046 | 92 | |
| BC067198.1 | - | 13 - 14 | 1581 | 6826682 | 92 | |
| U15635.1 | - | 13 - 14 | 1582 | 92 | ||
More Information
Set Membership: |
|

Samhd1
Genomic Map