Assay Details
| Gene Symbol: Pdzrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018884.2 | NP_061372.2 | 3 - 4 | 924 | 65 | |
| GenBank mRNA | AF127084.1 | - | 3 - 4 | 924 | 65 | |
| AF127085.1 | - | 4 - 5 | 768 | 65 | ||
| AK143226.1 | - | 3 - 4 | 438 | 65 | ||
| AK173098.1 | - | 3 - 4 | 883 | 65 | ||
| HM560981.1 | - | 3 - 4 | 915 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdzrn3
Genomic Map