Assay Details
| Gene Symbol: Vcan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081249.1 | NP_001074718.1 | 4 - 5 | 1065 | 81 | |
| NM_001134474.1 | NP_001127946.1 | 4 - 5 | 1065 | 81 | ||
| NM_001134475.1 | NP_001127947.1 | 4 - 5 | 1065 | 81 | ||
| NM_019389.2 | NP_062262.2 | 4 - 5 | 1065 | 81 | ||
| NM_172955.1 | NP_766543.1 | 4 - 5 | 1065 | 81 | ||
| GenBank mRNA | AK014525.1 | - | 4 - 5 | 875 | 81 | |
| AK034871.1 | - | 4 - 5 | 872 | 81 | ||
| AK040503.1 | - | 1 - 2 | 199 | 81 | ||
| AK135324.1 | - | 4 - 5 | 1062 | 81 | ||
| AK163927.1 | - | 4 - 5 | 922 | 81 | ||
| AK165069.1 | - | 4 - 5 | 868 | 81 | ||
| BC021652.1 | - | 4 - 5 | 905 | 81 | ||
| BC096495.1 | - | 4 - 5 | 875 | 81 | ||
| D16263.1 | - | 3 - 4 | 625 | 81 | ||
| D28599.1 | - | 4 - 5 | 797 | 81 | ||
| D32040.1 | - | 4 - 5 | 797 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vcan
Genomic Map