Assay Details
| Gene Symbol: Vcan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081249.1 | NP_001074718.1 | 5 - 6 | 1194 | 72 | |
| NM_001134474.1 | NP_001127946.1 | 5 - 6 | 1194 | 72 | ||
| NM_001134475.1 | NP_001127947.1 | 5 - 6 | 1194 | 72 | ||
| NM_019389.2 | NP_062262.2 | 5 - 6 | 1194 | 72 | ||
| NM_172955.1 | NP_766543.1 | 5 - 6 | 1194 | 72 | ||
| GenBank mRNA | AK014525.1 | - | 5 - 6 | 1004 | 72 | |
| AK034871.1 | - | 5 - 6 | 1001 | 72 | ||
| AK040503.1 | - | 2 - 3 | 328 | 72 | ||
| AK135324.1 | - | 5 - 6 | 1191 | 72 | ||
| AK163927.1 | - | 5 - 6 | 1051 | 72 | ||
| AK165069.1 | - | 5 - 6 | 997 | 72 | ||
| BC096495.1 | - | 5 - 6 | 1004 | 72 | ||
| D16263.1 | - | 4 - 5 | 754 | 72 | ||
| D28599.1 | - | 5 - 6 | 926 | 72 | ||
| D32040.1 | - | 5 - 6 | 926 | 72 | ||
More Information
Set Membership: |
|

Vcan
Genomic Map