Assay Details
| Gene Symbol: Sh3glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282037.1 | NP_001268966.1 | 9 - 10 | 1104 | 106 | |
| NM_001282042.1 | NP_001268971.1 | 7 - 8 | 1041 | 106 | ||
| NM_019464.3 | NP_062337.1 | 7 - 8 | 1041 | 106 | ||
| GenBank mRNA | AF263364.1 | - | 7 - 8 | 1029 | 106 | |
| AF272946.1 | - | 9 - 10 | 1092 | 106 | ||
| AK004590.1 | - | 7 - 8 | 852 | 106 | ||
| AK029647.1 | - | 6 - 7 | 702 | 106 | ||
| AK146246.1 | - | 7 - 8 | 870 | 106 | ||
| AK158415.1 | - | 7 - 8 | 817 | 106 | ||
| AK172959.1 | - | 7 - 8 | 852 | 106 | ||
| AK184553.1 | - | 1 - 2 | 107 | |||
| BC024362.1 | - | 7 - 8 | 851 | 5031296 | 106 | |
| DQ205686.1 | - | 7 - 8 | 1029 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3glb1
Genomic Map