Assay Details
| Gene Symbol: Crtam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019465.4 | NP_062338.3 | 7 - 8 | 867 | 69 | |
| GenBank mRNA | AF001104.1 | - | 7 - 8 | 813 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crtam
Genomic Map