Assay Details
| Gene Symbol: Acsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033600.1 | NP_001028772.1 | 10 - 11 | 1425 | 73 | |
| NM_019477.3 | NP_062350.3 | 10 - 11 | 1428 | 73 | ||
| NM_207625.2 | NP_997508.1 | 10 - 11 | 1718 | 73 | ||
| GenBank mRNA | AJ243502.1 | - | 10 - 11 | 1273 | 73 | |
| AK054387.1 | - | 10 - 11 | 1398 | 73 | ||
| BC016416.1 | - | 10 - 11 | 1299 | 3157957 | 73 | |
| BC058663.1 | - | 10 - 11 | 1514 | 6409748 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsl4
Genomic Map