Assay Details
| Gene Symbol: Rabggta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019519.2 | NP_062392.1 | 4 - 5 | 762 | 60 | |
| GenBank mRNA | AF127656.1 | - | 5 - 6 | 610 | 60 | |
| AF127657.1 | - | 4 - 5 | 553 | 60 | ||
| AF127658.1 | - | 4 - 5 | 762 | 60 | ||
| AF127659.1 | - | 4 - 5 | 762 | 60 | ||
| AF127660.1 | - | 5 - 6 | 610 | 60 | ||
| AF127661.1 | - | 4 - 5 | 553 | 60 | ||
| AF127662.1 | - | 5 - 6 | 681 | 60 | ||
| AK002625.1 | - | 5 - 6 | 544 | 60 | ||
| AK146917.1 | - | 5 - 6 | 633 | 60 | ||
| BC012214.1 | - | 5 - 6 | 615 | 3967218 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabggta
Genomic Map