Assay Details
| Gene Symbol: Rabggta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019519.2 | NP_062392.1 | 10 - 11 | 1385 | 79 | |
| GenBank mRNA | AF127656.1 | - | 11 - 12 | 1233 | 79 | |
| AF127657.1 | - | 10 - 11 | 1176 | 79 | ||
| AF127658.1 | - | 10 - 11 | 1385 | 79 | ||
| AF127659.1 | - | 10 - 11 | 1385 | 79 | ||
| AF127660.1 | - | 10 - 11 | 1330 | 79 | ||
| AF127661.1 | - | 9 - 10 | 1273 | 79 | ||
| AF127662.1 | - | 11 - 12 | 1304 | 79 | ||
| AK002625.1 | - | 11 - 12 | 1167 | 79 | ||
| AK146917.1 | - | 11 - 12 | 1256 | 79 | ||
| BC012214.1 | - | 9 - 10 | 1419 | 3967218 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabggta
Genomic Map