Assay Details
| Gene Symbol: Gas6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019521.2 | NP_062394.2 | 11 - 12 | 1500 | 67 | |
| GenBank mRNA | AK086187.1 | - | 11 - 12 | 1495 | 67 | |
| BC005444.1 | - | 11 - 12 | 1455 | 3592398 | 67 | |
| X59846.1 | - | 11 - 12 | 1471 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas6
Genomic Map