Assay Details
| Gene Symbol: Sh3gl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019535.2 | NP_062408.2 | 1 - 2 | 159 | 59 | |
| GenBank mRNA | AF326561.1 | - | 1 - 2 | 38 | 59 | |
| AF326562.1 | - | 1 - 2 | 38 | 59 | ||
| AK035210.1 | - | 490 | 59 | |||
| AK043424.1 | - | 147 | 59 | |||
| AK044370.1 | - | 1 - 2 | 174 | 59 | ||
| AK139337.1 | - | 1 - 2 | 124 | 59 | ||
| BC018385.1 | - | 1 - 2 | 159 | 4503038 | 59 | |
| U58886.1 | - | 1 - 2 | 122 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3gl2
Genomic Map