Assay Details
| Gene Symbol: Sh3gl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019535.2 | NP_062408.2 | 2 - 3 | 232 | 103 | |
| GenBank mRNA | AF326561.1 | - | 2 - 3 | 111 | 103 | |
| AF326562.1 | - | 2 - 3 | 111 | 103 | ||
| AK035210.1 | - | 563 | 103 | |||
| AK043424.1 | - | 220 | 103 | |||
| BC018385.1 | - | 2 - 3 | 232 | 4503038 | 103 | |
| U58886.1 | - | 2 - 3 | 195 | 103 | ||
More Information
Set Membership: |
|

Sh3gl2
Genomic Map